Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GEM
Entrez Gene ID
816780
Gene Name
GRAM domain family protein
Gene Aliases
AT2G22475, GL2-EXPRESSION MODULATOR
Chromosome Location
Chr.2: 9541128 - 9545038 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127813.3 NP_565538.1 1 - 2 735 110
NM_201781.1 NP_973510.1 1 - 2 462 110
GenBank mRNA EF490993.1 - 1 - 2 340 110

View Full Product Details