Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCPL9
Entrez Gene ID
816832
Gene Name
serine carboxypeptidase-like 9
Gene Aliases
AT2G23010, F21P24.7, F21P24_7, serine carboxypeptidase-like 9
Chromosome Location
Chr.2: 9798757 - 9802676 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127866.3 NP_179884.1 10 - 11 1035 68
NM_201788.2 NP_973517.1 10 - 11 981 68

View Full Product Details