Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLF
Entrez Gene ID
816870
Gene Name
SET domain-containing protein
Gene Aliases
AT2G23380, CURLY LEAF, F26B6.3, F26B6_3, ICU1, INCURVATA 1, SDG1, SET1, SETDOMAIN 1, SETDOMAIN GROUP 1
Chromosome Location
Chr.2: 9955092 - 9960386 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335847.1 NP_001324816.1 2 - 3 248 65
NM_127902.6 NP_179919.1 7 - 8 1373 65

View Full Product Details