Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl22
Entrez Gene ID
816934
Gene Name
serine carboxypeptidase-like 22
Gene Aliases
AT2G24000, T29E15.20, T29E15_20, serine carboxypeptidase-like 22
Chromosome Location
Chr.2: 10209421 - 10214042 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127962.3 NP_179978.2 3 - 4 511 125
GenBank mRNA DQ446558.1 - 3 - 4 457 125
DQ785742.1 - 3 - 4 457 125

View Full Product Details