Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPS10
Entrez Gene ID
816955
Gene Name
terpene synthase 10
Gene Aliases
AT2G24210, F27D4.12, F27D4_12, terpene synthase 10
Chromosome Location
Chr.2: 10294286 - 10297597 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127982.4 NP_179998.1 4 - 5 1120 143
GenBank mRNA BT033153.1 - 4 - 5 1076 143

View Full Product Details