Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFN3
Entrez Gene ID
831516
Gene Name
zinc finger nuclease 3
Gene Aliases
AT5G16540, MQK4.29, MQK4_29, zinc finger nuclease 3
Chromosome Location
Chr.5: 5403069 - 5405173 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343455.1 NP_001330309.1 3 - 4 867 148
NM_121660.4 NP_568332.2 4 - 5 635 148
NM_180710.3 NP_851041.1 4 - 5 769 148
NM_203061.2 NP_974790.1 3 - 4 867 148
GenBank mRNA AF138872.1 - 4 - 5 401 148
AY084634.1 - 4 - 5 558 148

View Full Product Details