Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCBK1
Entrez Gene ID
832411
Gene Name
long-chain base (LCB) kinase 1
Gene Aliases
AT5G23450, ATLCBK1, K19M13.8, K19M13_8, SPHINGOSINE KINASE, long-chain base (LCB) kinase 1
Chromosome Location
Chr.5: 7904293 - 7909623 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036852.4 NP_001031929.4 9 - 10 2637 96
NM_001343803.1 NP_001331260.1 8 - 9 2476 96
NM_001343804.1 NP_001331261.1 9 - 10 2458 96
NM_122252.4 NP_568432.1 8 - 9 2630 96
NM_180734.3 NP_851065.1 8 - 9 2496 96
GenBank mRNA AB046717.1 - 7 - 8 2136 96
AY139759.1 - 8 - 9 2552 96
BT004544.1 - 7 - 8 2136 96

View Full Product Details