Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ECT3
- Entrez Gene ID
-
836223
- Gene Name
- evolutionarily conserved C-terminal region 3
- Gene Aliases
-
AT5G61020, MAF19.3, MAF19_3, evolutionarily conserved C-terminal region 3
- Chromosome Location
-
Chr.5: 24556994 - 24559936 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_180905.3 |
NP_851236.1 |
2 - 3 |
248 |
61 |
View Full Product Details