Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chx28
Entrez Gene ID
824371
Gene Name
cation/hydrogen exchanger 28
Gene Aliases
AT3G52080, cation/hydrogen exchanger 28
Chromosome Location
Chr.3: 19314966 - 19317988 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115067.3 NP_190776.2 1 - 2 297 76

View Full Product Details