The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001339599.1 | NP_001325713.1 | 2 - 3 | 553 | 110 |
NM_115180.3 | NP_566980.1 | 3 - 4 | 465 | 110 | |
GenBank mRNA | AK117256.1 | - | 3 - 4 | 420 | 110 |
AY519599.1 | - | 3 - 4 | 352 | 110 | |
BT005257.1 | - | 3 - 4 | 352 | 110 |