Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE40
Entrez Gene ID
824537
Gene Name
Pentatricopeptide repeat (PPR) superfamily protein
Gene Aliases
AT3G53700, F5K20.2, maternal effect embryo arrest 40
Chromosome Location
Chr.3: 19900240 - 19903173 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115230.3 NP_190938.1 1 - 1 2516 70

View Full Product Details