Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HTA11
Entrez Gene ID
824621
Gene Name
histone H2A 11
Gene Aliases
AT3G54560, histone H2A 11
Chromosome Location
Chr.3: 20196140 - 20197726 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339683.1 NP_001325682.1 1 - 2 319 86
NM_115313.5 NP_191019.1 2 - 3 336 86
GenBank mRNA AK228050.1 - 2 - 3 206 86
AY087232.1 - 2 - 3 205 86
BT005205.1 - 1 - 2 116 86
Y12575.1 - - 214 86

View Full Product Details