Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATVPS33
Entrez Gene ID
824651
Gene Name
Sec1/munc18-like (SM) proteins superfamily
Gene Aliases
AT3G54860, VACUOLAR PROTEIN SORTING 33, VPS33
Chromosome Location
Chr.3: 20324088 - 20330030 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161207.1 NP_001154679.1 15 - 16 1248 120
NM_115343.4 NP_567009.1 14 - 15 1313 120
GenBank mRNA AF357527.1 - 14 - 15 1133 120

View Full Product Details