Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ROPGEF6
Entrez Gene ID
824732
Gene Name
ROP (rho of plants) guanine nucleotide exchange factor 6
Gene Aliases
AT3G55660, ATROPGEF6, KINASE PARTNER PROTEIN-LIKE, KPP-LIKE, ROP (rho of plants) guanine nucleotide exchange factor 6
Chromosome Location
Chr.3: 20648794 - 20651484 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339743.1 NP_001325478.1 2 - 3 591 76
NM_115424.3 NP_191125.1 2 - 3 632 76
GenBank mRNA BT020279.1 - 2 - 3 515 76
BT021117.1 - 2 - 3 438 76

View Full Product Details