Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SBPASE
Entrez Gene ID
824746
Gene Name
sedoheptulose-bisphosphatase
Gene Aliases
AT3G55800, sedoheptulose-bisphosphatase
Chromosome Location
Chr.3: 20709294 - 20711640 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115438.4 NP_191139.1 7 - 8 1243 86

View Full Product Details