Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACA11
Entrez Gene ID
824900
Gene Name
autoinhibited Ca2+-ATPase 11
Gene Aliases
AT3G57330, autoinhibited Ca2+-ATPase 11
Chromosome Location
Chr.3: 21211014 - 21216836 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115593.7 NP_191292.1 1 - 2 634 100

View Full Product Details