Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK32
Entrez Gene ID
824920
Gene Name
calcium-dependent protein kinase 32
Gene Aliases
AT3G57530, ATCPK32, CDPK32, calcium-dependent protein kinase 32
Chromosome Location
Chr.3: 21296457 - 21299704 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339876.1 NP_001326797.1 6 - 7 1884 106
NM_115613.4 NP_191312.2 7 - 8 1805 106
GenBank mRNA BT011630.1 - 7 - 8 1506 106
BT012274.1 - 7 - 8 1452 106

View Full Product Details