Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- UBP11
- Entrez Gene ID
-
840179
- Gene Name
- ubiquitin-specific protease 11
- Gene Aliases
-
AT1G32850, F9L11.5, F9L11_5, ubiquitin-specific protease 11
- Chromosome Location
-
Chr.1: 11902629 - 11907049 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103019.3 |
NP_174562.2 |
5 - 6 |
656 |
119 |
View Full Product Details