Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MES14
Entrez Gene ID
840296
Gene Name
methyl esterase 14
Gene Aliases
AT1G33990, ATMES14, F12G12.19, F12G12_19, METHYL ESTERASE 14, methyl esterase 14
Chromosome Location
Chr.1: 12355557 - 12358031 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103121.4 NP_174661.1 1 - 2 605 90
GenBank mRNA AF424607.1 - 1 - 2 594 90
AK226919.1 - 1 - 2 407 90
BT025240.1 - 1 - 2 253 90

View Full Product Details