Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP5
Entrez Gene ID
840329
Gene Name
receptor like protein 5
Gene Aliases
AT1G34290, AtRLP5, F23M19.6, F23M19_6, receptor like protein 5
Chromosome Location
Chr.1: 12498000 - 12498800 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103151.1 NP_174689.1 1 - 1 641 90

View Full Product Details