Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCB1;5
Entrez Gene ID
840348
Gene Name
CYCLIN B1;5
Gene Aliases
AT1G34460, CYC3, CYCLIN 3, CYCLIN B1;5, F12K21.22, F12K21_22
Chromosome Location
Chr.1: 12595110 - 12602379 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333104.1 NP_001319145.1 5 - 6 720 117
NM_001333106.1 NP_001320186.1 5 - 6 720 117
NM_001333108.1 NP_001320187.1 5 - 6 720 117
NM_001333109.1 NP_001320190.1 5 - 6 720 117
NM_001333110.1 NP_001320189.1 5 - 6 720 117
NM_001333111.1 NP_001320191.1 4 - 5 678 117
NM_103168.4 NP_564446.3 5 - 6 720 117

View Full Product Details