Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRF11
Entrez Gene ID
840380
Gene Name
general regulatory factor 11
Gene Aliases
14-3-3 PROTEIN GF14OMICRON, AT1G34760, F11O6.13, GF14 OMICRON, RHS5, ROOT HAIR SPECIFIC 5, general regulatory factor 11
Chromosome Location
Chr.1: 12743722 - 12745748 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084180.2 NP_001077649.1 4 - 5 718 89
NM_103196.4 NP_564451.2 4 - 5 718 89
GenBank mRNA BT029457.1 - 4 - 5 573 89

View Full Product Details