Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLO9
Entrez Gene ID
840861
Gene Name
Seven transmembrane MLO family protein
Gene Aliases
ARABIDOPSIS THALIANA MILDEW RESISTANCE LOCUS O 9, AT1G42560, ATMLO9, MILDEW RESISTANCE LOCUS O 9, T8D8.5, T8D8_5
Chromosome Location
Chr.1: 15980135 - 15983571 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333203.1 NP_001322027.1 8 - 9 1021 86
NM_103440.4 NP_174980.3 8 - 9 1104 86

View Full Product Details