Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RP1
Entrez Gene ID
840916
Gene Name
ribosomal protein 1
Gene Aliases
ARP1, AT1G43170, F1I21.1, F1I21_1, RPL3A, emb2207, embryo defective 2207, ribosomal protein 1
Chromosome Location
Chr.1: 16266553 - 16268945 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036069.1 NP_001031146.1 4 - 5 534 71
NM_001084202.1 NP_001077671.1 2 - 3 360 71
NM_001198219.1 NP_001185148.1 4 - 5 650 71
NM_001198220.1 NP_001185149.1 4 - 5 653 71
NM_001198221.1 NP_001185150.1 4 - 5 659 71
NM_001198222.1 NP_001185151.1 4 - 5 656 71
NM_001198223.1 NP_001185152.1 4 - 5 662 71
NM_103469.4 NP_175009.1 4 - 5 750 71
NM_202237.1 NP_973966.1 4 - 5 536 71
GenBank mRNA AF327421.1 - 4 - 5 559 71
AF361101.1 - 3 - 4 496 71
AF361809.1 - 4 - 5 555 71
AX765804.1 - 3 - 4 496 71
AY039544.1 - 4 - 5 559 71
AY052743.1 - 3 - 4 496 71
AY057487.1 - 4 - 5 553 71

View Full Product Details