Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGAL5
Entrez Gene ID
841080
Gene Name
beta-galactosidase 5
Gene Aliases
AT1G45130, AtBGAL5, F27F5.20, F27F5_20, beta-galactosidase 5
Chromosome Location
Chr.1: 17065111 - 17069269 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333243.1 NP_001320473.1 14 - 15 1885 79
NM_103587.3 NP_175127.1 16 - 17 2417 79
GenBank mRNA AY069911.1 - 16 - 17 2216 79
AY093977.1 - 16 - 17 2081 79

View Full Product Details