Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VFB1
Entrez Gene ID
841121
Gene Name
VIER F-box protein 1
Gene Aliases
AT1G47056, F2G19.16, F2G19_16, VIER F-box proteine 1
Chromosome Location
Chr.1: 17275890 - 17277839 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103611.2 NP_175151.1 1 - 1 1679 81

View Full Product Details