Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KU80
Entrez Gene ID
841223
Gene Name
Ku80 family protein
Gene Aliases
ARABIDOPSIS THALIANA KU80 HOMOLOG, AT1G48050, ATKU80, KU80-LIKE PROTEIN, T2J15.4
Chromosome Location
Chr.1: 17723382 - 17727243 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333292.1 NP_001322094.1 5 - 6 767 71
NM_001333293.1 NP_001322093.1 4 - 5 695 71
NM_103701.2 NP_564520.1 5 - 6 815 71
GenBank mRNA AF283758.1 - 5 - 6 745 71

View Full Product Details