Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KU80
Entrez Gene ID
841223
Gene Name
Ku80 family protein
Gene Aliases
ARABIDOPSIS THALIANA KU80 HOMOLOG, AT1G48050, ATKU80, KU80-LIKE PROTEIN, T2J15.4
Chromosome Location
Chr.1: 17723382 - 17727243 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333292.1 NP_001322094.1 8 - 9 1455 90
NM_001333293.1 NP_001322093.1 7 - 8 1383 90
NM_103701.2 NP_564520.1 8 - 9 1503 90
GenBank mRNA AF283758.1 - 8 - 9 1433 90

View Full Product Details