Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DL1E
Entrez Gene ID
825189
Gene Name
DYNAMIN-like 1E
Gene Aliases
ADL1E, ADL4, ADLP2, ARABIDOPSIS DYNAMIN-LIKE 4, AT3G60190, DRP1E, DYNAMIN-LIKE PROTEIN 2, DYNAMIN-RELATED PROTEIN 1E, DYNAMIN-like 1E, EDR3, ENHANCED DISEASE RESISTANCE 3, F27H5.4
Chromosome Location
Chr.3: 22244169 - 22247976 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115882.4 NP_567094.1 3 - 4 641 141
GenBank mRNA AF428332.1 - 3 - 4 463 141
AJ304842.1 - 3 - 4 423 141

View Full Product Details