Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG8G
Entrez Gene ID
825235
Gene Name
Ubiquitin-like superfamily protein
Gene Aliases
AT3G60640, AUTOPHAGY 8G
Chromosome Location
Chr.3: 22415751 - 22417548 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115928.5 NP_191623.1 2 - 3 211 86
GenBank mRNA AB073181.1 - 2 - 3 98 86
AY062844.1 - 2 - 3 121 86
AY088249.1 - 2 - 3 121 86
BT001196.1 - 1 - 2 37 86

View Full Product Details