Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAD3
Entrez Gene ID
817548
Gene Name
fatty acid desaturase 3
Gene Aliases
AT2G29980, AtFAD3, ENDOPLASMIC RETICULUM 18:2 DESATURASE, F23F1.10, F23F1_10, OMEGA-3 FATTY ACID DESATURASE, fatty acid desaturase 3
Chromosome Location
Chr.2: 12781524 - 12785190 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128552.4 NP_180559.1 3 - 4 709 86
NM_179808.2 NP_850139.1 3 - 4 556 86
GenBank mRNA AY063966.1 - 3 - 4 571 86
AY096462.1 - 3 - 4 465 86
D17579.1 - 3 - 4 547 86

View Full Product Details