Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABO1
- Entrez Gene ID
-
831213
- Gene Name
- IKI3 family protein
- Gene Aliases
-
ABA-OVERLY SENSITIVE 1, AT5G13680, AtELP1, ELO2, ELONGATA 2, MSH12.15, MSH12_15
- Chromosome Location
-
Chr.5: 4410314 - 4415622 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121371.3 |
NP_196872.1 |
4 - 5 |
3794 |
87 |
View Full Product Details