Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHB5
Entrez Gene ID
831280
Gene Name
prohibitin 5
Gene Aliases
AT5G14300, ATPHB5, F18O22.90, F18O22_90, prohibitin 5
Chromosome Location
Chr.5: 4613102 - 4614023 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121434.1 NP_196934.1 1 - 2 286 80

View Full Product Details