Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IRX6
Entrez Gene ID
831417
Gene Name
COBRA-like extracellular glycosyl-phosphatidyl inositol-anchored protein family
Gene Aliases
AT5G15630, COBL4, COBRA-LIKE4, F14F8.10, F14F8_10, IRREGULAR XYLEM 6
Chromosome Location
Chr.5: 5084524 - 5086811 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343411.1 NP_001331539.1 2 - 3 1142 75
NM_001343412.1 NP_001331538.1 3 - 4 1051 75
NM_001343413.1 NP_001331540.1 3 - 4 1033 75
NM_001343414.1 NP_001331537.1 4 - 5 886 75
NM_121567.5 NP_197067.2 4 - 5 1057 75
GenBank mRNA BT014900.1 - 4 - 5 921 75
BT020445.1 - 4 - 5 739 75

View Full Product Details