Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TERT
Entrez Gene ID
831548
Gene Name
telomerase reverse transcriptase
Gene Aliases
AT5G16850, ATTERT, F5E19.190, F5E19_190, telomerase reverse transcriptase
Chromosome Location
Chr.5: 5538180 - 5543657 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121691.4 NP_197187.1 10 - 11 2943 104
GenBank mRNA AF135454.1 - 10 - 11 2927 104

View Full Product Details