Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DAR4
- Entrez Gene ID
-
831657
- Gene Name
- DA1-related protein 4
- Gene Aliases
-
AT5G17890, CHILLING SENSITIVE 3, CHS3, DA1-related protein 4, MPI7.6, MPI7_6
- Chromosome Location
-
Chr.5: 5916862 - 5923494 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121795.4 |
NP_197291.2 |
8 - 9 |
4184 |
75 |
View Full Product Details