Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FASS
Entrez Gene ID
831976
Gene Name
tonneau 2 (TON2)
Gene Aliases
AT5G18580, EMB40, EMBRYO DEFECTIVE 40, FASS 1, FASS 2, FS1, GDO, GORDO, T28N17.60, T28N17_60, TON2, TONNEAU 2
Chromosome Location
Chr.5: 6174972 - 6178401 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121863.4 NP_568364.1 5 - 6 740 92
GenBank mRNA AF280057.1 - 5 - 6 716 92
AF360316.1 - 5 - 6 703 92
AY113888.1 - 5 - 6 558 92

View Full Product Details