Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT20
Entrez Gene ID
832066
Gene Name
nudix hydrolase homolog 20
Gene Aliases
AT5G19460, F7K24.210, F7K24_210, atnudt20, nudix hydrolase homolog 20
Chromosome Location
Chr.5: 6562749 - 6565720 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121951.5 NP_197447.2 4 - 5 975 124
GenBank mRNA AY074342.1 - 4 - 5 625 124
AY117160.1 - 4 - 5 612 124

View Full Product Details