Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C4H
Entrez Gene ID
817599
Gene Name
cinnamate-4-hydroxylase
Gene Aliases
AT2G30490, ATC4H, CINNAMATE 4-HYDROXYLASE, CINNAMATE-4-HYDROXYLASE, CYP73A5, REDUCED EPRDERMAL FLUORESCENCE 3, REF3, T6B20.16, T6B20_16, cinnamate-4-hydroxylase
Chromosome Location
Chr.2: 12993625 - 12996012 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128601.3 NP_180607.1 2 - 3 1250 87
GenBank mRNA AY065145.1 - 2 - 3 1008 87
BT008875.1 - 2 - 3 921 87
U71081.1 - 2 - 3 976 87

View Full Product Details