Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- OFP16
- Entrez Gene ID
-
817770
- Gene Name
- ovate family protein 16
- Gene Aliases
-
AT2G32100, ATOFP16, F22D22.15, F22D22_15, RABIDOPSIS THALIANA OVATE FAMILY PROTEIN 16, ovate family protein 16
- Chromosome Location
-
Chr.2: 13647699 - 13649002 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128770.3 |
NP_180770.1 |
1 - 1 |
900 |
71 |
View Full Product Details