The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_128785.3 | NP_180785.1 | 1 - 2 | 372 | 79 |
GenBank mRNA | AY136393.1 | - | 1 - 2 | 305 | 79 |
BT000207.1 | - | 1 - 2 | 184 | 79 |