Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCT1
Entrez Gene ID
817786
Gene Name
phosphorylcholine cytidylyltransferase
Gene Aliases
AT2G32260, ATCCT1, T32F6.22, T32F6_22, phosphorylcholine cytidylyltransferase
Chromosome Location
Chr.2: 13697457 - 13700648 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336383.1 NP_001323485.1 6 - 7 1358 102
NM_128785.3 NP_180785.1 7 - 8 956 102
GenBank mRNA AY136393.1 - 7 - 8 889 102
BT000207.1 - 7 - 8 768 102

View Full Product Details