Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP24
Entrez Gene ID
817864
Gene Name
receptor like protein 24
Gene Aliases
AT2G33020, AtRLP24, T21L14.1, receptor like protein 24
Chromosome Location
Chr.2: 14013797 - 14016516 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128862.2 NP_180861.1 1 - 2 411 100

View Full Product Details