Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RALFL18
Entrez Gene ID
817875
Gene Name
ralf-like 18
Gene Aliases
AT2G33130, F25I18.13, F25I18_13, RALF18, ralf-like 18
Chromosome Location
Chr.2: 14046388 - 14047063 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128873.2 NP_180872.1 1 - 1 282 71

View Full Product Details