Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CUTA
Entrez Gene ID
817940
Gene Name
Nitrogen regulatory PII-like, alpha/beta
Gene Aliases
AT2G33740, T1B8.5, T1B8_5
Chromosome Location
Chr.2: 14269461 - 14271723 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128933.4 NP_180930.1 2 - 3 405 76
NM_179886.1 NP_850217.1 2 - 3 285 76
GenBank mRNA AF327524.1 - 2 - 3 276 76
AF327525.1 - 2 - 3 296 76
BT026102.1 - 2 - 3 276 76

View Full Product Details