Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATHB-15
Entrez Gene ID
841645
Gene Name
Homeobox-leucine zipper family protein / lipid-binding START domain-containing protein
Gene Aliases
AT1G52150, ATHB15, CNA, CORONA, F5F19.21, F5F19_21, ICU4, INCURVATA 4
Chromosome Location
Chr.1: 19409688 - 19414764 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084233.2 NP_001077702.1 1 - 2 1017 72
NM_104096.3 NP_175627.1 1 - 2 1017 72
NM_179464.1 NP_849795.1 1 - 2 931 72
GenBank mRNA AJ439449.1 - 1 - 2 214 72
AY060556.1 - 1 - 2 931 72

View Full Product Details