Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RR20
Entrez Gene ID
825441
Gene Name
two-component response regulator
Gene Aliases
ARR20, AT3G62670, MEE41, maternal effect embryo arrest 41, response regulator 20
Chromosome Location
Chr.3: 23176556 - 23178294 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340168.1 NP_001319821.1 2 - 3 337 91

View Full Product Details