Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATGSTF13
Entrez Gene ID
825451
Gene Name
Glutathione S-transferase family protein
Gene Aliases
AT3G62760
Chromosome Location
Chr.3: 23217233 - 23218312 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116141.2 NP_191835.1 2 - 3 267 90

View Full Product Details