Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRF
Entrez Gene ID
825494
Gene Name
ribosome recycling factor, chloroplast precursor
Gene Aliases
''ribosome recycling factor, AT3G63190, Arabidopsis thaliana chloroplast ribosome recycling factor, AtcpRRF, HFP108, HIGH CHLOROPHYLL FLUORESCENCE AND PALE GREEN MUTANT 108, chloroplast precursor, chloroplast precursor'', chloroplast ribosome recycling factor, cpRRF, ribosome recycling factor
Chromosome Location
Chr.3: 23342550 - 23344801 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116184.4 NP_567141.1 1 - 2 248 82
GenBank mRNA AF372883.1 - 1 - 2 136 82
AY056778.1 - 1 - 2 216 82
AY092982.1 - 1 - 2 154 82
BT002678.1 - 1 - 2 87 82

View Full Product Details