Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAM2
Entrez Gene ID
827959
Gene Name
beta-amylase 2
Gene Aliases
AT4G00490, BETA-AMYLASE 9, BMY9, F6N23.1, F6N23_1, beta-amylase 2
Chromosome Location
Chr.4: 222289 - 225131 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116273.5 NP_191958.3 4 - 5 835 85

View Full Product Details